siRNA Details
| Gene Symbol: Fgl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053455.2 | NP_445907.2 | 1 | 331 | ||
| GenBank mRNA | AF323608.2 | - | 1 | 358 | ||
| BC072502.1 | - | 1 | 280 | |||
| BC085357.1 | - | 1 | 288 | |||
| BC107665.1 | - | 1 | 304 | |||
| CB582252.1 | - | 1 | 342 | |||

Fgl2
Genomic Map