siRNA Details
| Gene Symbol: Nucb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053463.1 | NP_445915.1 | 2 | 175 | ||
| XM_006229163.1 | XP_006229225.1 | 3 | 292 | |||
| XM_006229164.2 | XP_006229226.1 | 3 | 236 | |||
| XM_006229165.1 | XP_006229227.1 | 3 | 292 | |||
| GenBank mRNA | AF336828.1 | - | 2 | 175 | ||
| BC062084.1 | - | 3 | 236 | |||
| BC100643.1 | - | 3 | 225 | 7930904 | ||
| Z36277.1 | - | 3 | 237 | |||

Nucb1
Genomic Map