siRNA Details
| Gene Symbol: Plg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053491.2 | NP_445943.1 | 2 | 175 | ||
| GenBank mRNA | AJ242649.1 | - | 2 | 178 | ||
| BC091135.1 | - | 2 | 175 | 7369357 | ||
| FQ210878.1 | - | 2 | 318 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Plg| Gene Symbol: Plg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053491.2 | NP_445943.1 | 2 | 175 | ||
| GenBank mRNA | AJ242649.1 | - | 2 | 178 | ||
| BC091135.1 | - | 2 | 175 | 7369357 | ||
| FQ210878.1 | - | 2 | 318 | |||