siRNA Details
| Gene Symbol: Cnga3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053495.1 | NP_445947.1 | 2 | 104 | ||
| XM_008767042.1 | XP_008765264.1 | 4 | 628 | |||
| XM_008767043.1 | XP_008765265.1 | 3 | 514 | |||
| XM_008767044.1 | XP_008765266.1 | 3 | 514 | |||
| GenBank mRNA | AB002801.1 | - | 2 | 104 | ||
| AJ272428.1 | - | 4 | 391 | |||
| AJ272429.1 | - | 3 | 277 | |||

Cnga3
Genomic Map