siRNA Details
| Gene Symbol: Nox4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053524.1 | NP_445976.1 | 2 | 175 | ||
| XM_008759640.1 | XP_008757862.1 | 2 | 93 | |||
| XM_008759641.1 | XP_008757863.1 | 2 | 92 | |||
| XM_008759642.1 | XP_008757864.1 | 1 | 69 | |||
| XM_008759643.1 | XP_008757865.1 | 2 | 298 | |||
| GenBank mRNA | AB044086.1 | - | 2 | 143 | ||
| AY027527.1 | - | 2 | 175 | |||

Nox4
Genomic Map