siRNA Details
| Gene Symbol: Cdc5l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053527.1 | NP_445979.1 | 3 | 313 | ||
| GenBank mRNA | AF000578.2 | - | 3 | 313 | ||
| BC087007.1 | - | 3 | 250 | |||
| BC161839.1 | - | 3 | 277 | 9101355 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cdc5l| Gene Symbol: Cdc5l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053527.1 | NP_445979.1 | 3 | 313 | ||
| GenBank mRNA | AF000578.2 | - | 3 | 313 | ||
| BC087007.1 | - | 3 | 250 | |||
| BC161839.1 | - | 3 | 277 | 9101355 | ||