siRNA Details
| Gene Symbol: Polg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053528.1 | NP_445980.1 | 2 | 734 | ||
| XM_006229410.2 | XP_006229472.1 | 3 | 1075 | |||
| XM_008759561.1 | XP_008757783.1 | 3 | 1077 | |||
| XM_008759562.1 | XP_008757784.1 | 3 | 1021 | |||
| GenBank mRNA | AJ245646.1 | - | 2 | 734 | ||
| AJ245647.1 | - | 2 | 734 | |||

Polg
Genomic Map