siRNA Details
| Gene Symbol: Twist1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053530.2 | NP_445982.1 | 1 | 650 | ||
| GenBank mRNA | AF266260.1 | - | 1 | 145 | ||
| BC166412.1 | - | 1 | 780 | 9033082 | ||
UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Gene Symbol: Twist1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053530.2 | NP_445982.1 | 1 | 650 | ||
| GenBank mRNA | AF266260.1 | - | 1 | 145 | ||
| BC166412.1 | - | 1 | 780 | 9033082 | ||
Twist1
Genomic Map