siRNA Details
| Gene Symbol: Gab2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053417.1 | NP_445869.1 | 2 | 257 | ||
| XM_008759806.1 | XP_008758028.1 | 1 | 1889 | |||
| GenBank mRNA | AF230367.2 | - | 2 | 257 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gab2| Gene Symbol: Gab2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053417.1 | NP_445869.1 | 2 | 257 | ||
| XM_008759806.1 | XP_008758028.1 | 1 | 1889 | |||
| GenBank mRNA | AF230367.2 | - | 2 | 257 | ||