siRNA Details
| Gene Symbol: Arid4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053421.1 | NP_445873.1 | 3 | 202 | ||
| XM_006254016.2 | XP_006254078.1 | 4 | 527 | |||
| XM_006254019.2 | XP_006254081.1 | 4 | 527 | |||
| XM_006254020.2 | XP_006254082.1 | 4 | 527 | |||
| XM_008771764.1 | XP_008769986.1 | 4 | 527 | |||
| XM_008771765.1 | XP_008769987.1 | 4 | 527 | |||
| XM_008771766.1 | XP_008769988.1 | 4 | 527 | |||
| GenBank mRNA | AF245512.1 | - | 3 | 202 | ||
| Gene Symbol: LOC100912163 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008771836.1 | XP_008770058.1 | 3 | 287 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_194262, NM_198122, AK136105, AK161669, BY720039, and others.

Arid4b
Genomic Map