siRNA Details
| Gene Symbol: Qsox1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109898.1 | NP_001103368.1 | 2, 3 | 409 | ||
| NM_053431.3 | NP_445883.1 | 2, 3 | 409 | |||
| GenBank mRNA | AB044285.1 | - | 2, 3 | 409 | ||
| AF217799.1 | - | 2, 3 | 413 | |||
| AF285078.1 | - | 2, 3 | 431 | |||
| AY623665.1 | - | 2, 3 | 411 | |||

Qsox1
Genomic Map