siRNA Details
| Gene Symbol: Rnf103 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053438.2 | NP_445890.2 | 2 | 278 | ||
| XM_006236653.2 | XP_006236715.1 | 2 | 1016 | |||
| GenBank mRNA | AF306394.1 | - | 2 | 1002 | ||
| BC100629.1 | - | 2 | 278 | 7938106 | ||

Rnf103
Genomic Map