siRNA Details
| Gene Symbol: Rgs2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053453.2 | NP_445905.1 | 4 | 406 | ||
| GenBank mRNA | AF279918.1 | - | 419 | |||
| AF321837.1 | - | 4 | 406 | |||
| AY043246.1 | - | 430 | ||||
| BC061969.1 | - | 427 | 5621945 | |||
| FQ224353.1 | - | 437 | ||||
| FQ228949.1 | - | 437 | ||||

Rgs2
Genomic Map