siRNA Details
| Gene Symbol: LOC100912604 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003749458.2 | XP_003749506.1 | 3, 4 | 448 | ||
| Gene Symbol: Srm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053464.2 | NP_445916.1 | 3, 4 | 451 | ||
| GenBank mRNA | AF337636.1 | - | 3, 4 | 454 | ||
| BC128733.1 | - | 3, 4 | 497 | 8366755 | ||

LOC100912604
Genomic Map