siRNA Details
| Gene Symbol: Slc5a7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053521.1 | NP_445973.1 | 3 | 427 | ||
| XM_006244251.2 | XP_006244313.1 | 3 | 530 | |||
| XM_006244253.2 | XP_006244315.1 | 1 | 74 | |||
| XM_008766774.1 | XP_008764996.1 | 3 | 578 | |||
| XM_008766775.1 | XP_008764997.1 | 3 | 527 | |||
| GenBank mRNA | AB030947.1 | - | 3 | 427 | ||

Slc5a7
Genomic Map