siRNA Details
| Gene Symbol: Ciita | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270803.1 | NP_001257732.1 | 3 | 351 | ||
| NM_001270804.1 | NP_001257733.1 | 3 | 279 | |||
| NM_053529.2 | NP_445981.2 | 3 | 570 | |||
| XM_006245738.2 | XP_006245800.1 | 3 | 612 | |||
| XM_008767501.1 | XP_008765723.1 | 4 | 1652 | |||
| GenBank mRNA | AF251305.1 | - | 3 | 570 | ||
| AF251306.1 | - | 3 | 351 | |||
| AF251307.1 | - | 3 | 279 | |||

Ciita
Genomic Map