siRNA Details
| Gene Symbol: Tle3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053400.1 | NP_445852.1 | 15 | 2242 | ||
| XM_006243211.2 | XP_006243273.1 | 16 | 2489 | |||
| XM_006243212.2 | XP_006243274.1 | 15 | 2662 | |||
| XM_006243213.2 | XP_006243275.1 | 15 | 2655 | |||
| XM_006243214.2 | XP_006243276.1 | 15 | 2650 | |||
| XM_006243215.2 | XP_006243277.1 | 15 | 2640 | |||
| XM_006243216.2 | XP_006243278.1 | 15 | 2634 | |||
| XM_006243217.2 | XP_006243279.1 | 15 | 2631 | |||
| XM_006243218.2 | XP_006243280.1 | 15 | 2624 | |||
| XM_006243219.2 | XP_006243281.1 | 15 | 2619 | |||
| XM_006243220.2 | XP_006243282.1 | 15 | 2616 | |||
| XM_006243221.2 | XP_006243283.1 | 15 | 2607 | |||
| XM_008766347.1 | XP_008764569.1 | 15 | 2664 | |||
| GenBank mRNA | AF186092.1 | - | 15 | 2242 | ||
| BC097419.1 | - | 15 | 1738 | 7454629 | ||

Tle3
Genomic Map