siRNA Details
| Gene Symbol: Dctn4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053404.2 | NP_445856.1 | 8 | 665 | ||
| XM_006254783.2 | XP_006254845.1 | 7 | 707 | |||
| GenBank mRNA | AF190798.1 | - | 8 | 726 | ||
| AF192493.1 | - | 7 | 644 | |||
| AF192494.1 | - | 8 | 665 | |||
| BC100143.1 | - | 7 | 649 | 7133708 | ||

Dctn4
Genomic Map