siRNA Details
| Gene Symbol: TESMIN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004923.3 | NP_004914.2 | 10 | 2321 | ||
| GenBank mRNA | BC064579.1 | - | 10 | 2288 | 5743865 | |
| U86074.1 | - | 10 | 1899 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TESMIN| Gene Symbol: TESMIN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004923.3 | NP_004914.2 | 10 | 2321 | ||
| GenBank mRNA | BC064579.1 | - | 10 | 2288 | 5743865 | |
| U86074.1 | - | 10 | 1899 | |||