siRNA Details
| Gene Symbol: Rab27b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053459.1 | NP_445911.1 | 5 | 593 | ||
| XM_006254930.2 | XP_006254992.1 | 6 | 555 | |||
| GenBank mRNA | AF325693.1 | - | 5 | 593 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rab27b| Gene Symbol: Rab27b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053459.1 | NP_445911.1 | 5 | 593 | ||
| XM_006254930.2 | XP_006254992.1 | 6 | 555 | |||
| GenBank mRNA | AF325693.1 | - | 5 | 593 | ||