siRNA Details
| Gene Symbol: Slc44a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033852.1 | NP_001029024.1 | 11 | 1388 | ||
| NM_053492.3 | NP_445944.2 | 11 | 1388 | |||
| XM_008763716.1 | XP_008761938.1 | 11 | 1421 | |||
| XM_008763717.1 | XP_008761939.1 | 11 | 1422 | |||
| GenBank mRNA | AJ245619.1 | - | 11 | 1380 | ||
| AJ420809.1 | - | 11 | 1388 | |||

Slc44a1
Genomic Map