siRNA Details
| Gene Symbol: Slc25a14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053501.2 | NP_445953.1 | 7 | 719 | ||
| XM_006257531.2 | XP_006257593.1 | 6 | 712 | |||
| XM_006257532.1 | XP_006257594.1 | 7 | 756 | |||
| XM_006257534.2 | XP_006257596.1 | 7 | 1652 | |||
| XM_006257535.1 | XP_006257597.1 | 7 | 574 | |||
| XM_008773530.1 | XP_008771752.1 | 6 | 717 | |||
| XM_008773531.1 | XP_008771753.1 | 8 | 609 | |||
| GenBank mRNA | AB028879.1 | - | 6 | 516 | ||
| AF300424.1 | - | 7 | 720 | |||
| AJ300165.1 | - | 6 | 525 | |||

Slc25a14
Genomic Map