siRNA Details
| Gene Symbol: Cpn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053526.2 | NP_445978.1 | 8 | 1351 | ||
| GenBank mRNA | AB042599.1 | - | 8 | 1360 | ||
| BC088124.1 | - | 8 | 1351 | 7326031 | ||
| FQ218535.1 | - | 3 | 165 | |||
| FQ219519.1 | - | 6 | 636 | |||

Cpn1
Genomic Map