siRNA Details
| Gene Symbol: Klf15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053536.1 | NP_445988.1 | 3 | 505 | ||
| XM_006236882.2 | XP_006236944.1 | 2 | 305 | |||
| XM_006236884.2 | XP_006236946.1 | 2 | 501 | |||
| XM_008763135.1 | XP_008761357.1 | 2 | 642 | |||
| XM_008763136.1 | XP_008761358.1 | 2 | 1464 | |||
| GenBank mRNA | AB020759.1 | - | 3 | 505 | ||
| AB102791.1 | - | 1 | 227 | |||
| AB102792.1 | - | 1 | 227 | |||
| BC089782.1 | - | 2 | 404 | 7325036 | ||

Klf15
Genomic Map