siRNA Details
| Gene Symbol: Angpt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053546.1 | NP_445998.1 | 1 | 387 | ||
| XM_006241609.2 | XP_006241671.1 | 1 | 729 | |||
| GenBank mRNA | AB080023.1 | - | 1 | 387 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Angpt1| Gene Symbol: Angpt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053546.1 | NP_445998.1 | 1 | 387 | ||
| XM_006241609.2 | XP_006241671.1 | 1 | 729 | |||
| GenBank mRNA | AB080023.1 | - | 1 | 387 | ||