siRNA Details
| Gene Symbol: Rpe65 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053562.3 | NP_446014.2 | 3 | 197 | ||
| GenBank mRNA | AF035673.1 | - | 3 | 199 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rpe65| Gene Symbol: Rpe65 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053562.3 | NP_446014.2 | 3 | 197 | ||
| GenBank mRNA | AF035673.1 | - | 3 | 199 | ||