siRNA Details
| Gene Symbol: Madd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053585.1 | NP_446037.1 | 3, 4 | 845 | ||
| GenBank mRNA | U72995.1 | - | 3, 4 | 845 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Madd| Gene Symbol: Madd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053585.1 | NP_446037.1 | 3, 4 | 845 | ||
| GenBank mRNA | U72995.1 | - | 3, 4 | 845 | ||