siRNA Details
| Gene Symbol: Csnk1a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053615.1 | NP_446067.1 | 1, 2 | 131 | ||
| GenBank mRNA | BC127486.1 | - | 357 | 8364080 | ||
| U77582.1 | - | 1, 2 | 131 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Csnk1a1| Gene Symbol: Csnk1a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053615.1 | NP_446067.1 | 1, 2 | 131 | ||
| GenBank mRNA | BC127486.1 | - | 357 | 8364080 | ||
| U77582.1 | - | 1, 2 | 131 | |||