siRNA Details
| Gene Symbol: Asah2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053646.1 | NP_446098.1 | 3 | 210 | ||
| XM_006231261.2 | XP_006231323.1 | 6 | 568 | |||
| XM_006231263.1 | XP_006231325.1 | 5 | 1146 | |||
| XM_006231267.2 | XP_006231329.1 | 5 | 1152 | |||
| XM_006231268.2 | XP_006231330.1 | 5 | 1149 | |||
| XM_006231270.1 | XP_006231332.1 | 5 | 1149 | |||
| XM_008760319.1 | XP_008758541.1 | 8 | 2988 | |||
| XM_008760320.1 | XP_008758542.1 | 5 | 429 | |||
| GenBank mRNA | AB057433.1 | - | 3 | 210 | ||

Asah2
Genomic Map