siRNA Details
| Gene Symbol: Cxcl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053647.1 | NP_446099.1 | 2 | 209 | ||
| GenBank mRNA | AB060092.1 | - | 2 | 209 | ||
| S77604.1 | - | 2 | 164 | |||
| U45965.1 | - | 2 | 172 | |||
| X65647.1 | - | 2 | 149 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cxcl2| Gene Symbol: Cxcl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053647.1 | NP_446099.1 | 2 | 209 | ||
| GenBank mRNA | AB060092.1 | - | 2 | 209 | ||
| S77604.1 | - | 2 | 164 | |||
| U45965.1 | - | 2 | 172 | |||
| X65647.1 | - | 2 | 149 | |||