siRNA Details
| Gene Symbol: Sec16b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053571.1 | NP_446023.1 | 2 | 387 | ||
| XM_008769682.1 | XP_008767904.1 | 8 | 3919 | |||
| XM_008769683.1 | XP_008767905.1 | 6 | 3627 | |||
| XM_008769684.1 | XP_008767906.1 | 3 | 1151 | |||
| XM_008769685.1 | XP_008767907.1 | 5 | 2904 | |||
| XM_008769686.1 | XP_008767908.1 | 1 | 181 | |||
| GenBank mRNA | AB060653.1 | - | 2 | 387 | ||

Sec16b
Genomic Map