siRNA Details
| Gene Symbol: Tinagl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053582.2 | NP_446034.1 | 7 | 1005 | ||
| GenBank mRNA | AB050717.1 | - | 7 | 1006 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tinagl1| Gene Symbol: Tinagl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053582.2 | NP_446034.1 | 7 | 1005 | ||
| GenBank mRNA | AB050717.1 | - | 7 | 1006 | ||