siRNA Details
| Gene Symbol: Nupr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053611.2 | NP_446063.1 | 2 | 285 | ||
| GenBank mRNA | AF014503.1 | - | 2 | 253 | ||
| Gene Symbol: NEWGENE_69363 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008759924.1 | XP_008758146.1 | 2 | 336 | ||

Nupr1
Genomic Map