siRNA Details
| Gene Symbol: Csnk1a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053615.1 | NP_446067.1 | 3 | 252 | ||
| GenBank mRNA | U77582.1 | - | 3 | 252 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Csnk1a1| Gene Symbol: Csnk1a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053615.1 | NP_446067.1 | 3 | 252 | ||
| GenBank mRNA | U77582.1 | - | 3 | 252 | ||