siRNA Details
| Gene Symbol: Cpb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053617.2 | NP_446069.1 | 3 | 238 | ||
| GenBank mRNA | BC091133.1 | - | 3 | 202 | ||
| BC107447.1 | - | 3 | 231 | 7323668 | ||
| DY312674.1 | - | 3 | 238 | |||
| FQ218610.1 | - | 3 | 243 | |||
| FQ218635.1 | - | 3 | 192 | |||
| FQ218700.1 | - | 3 | 243 | |||
| FQ219505.1 | - | 1 | 24 | |||
| FQ219645.1 | - | 3 | 310 | |||

Cpb2
Genomic Map