siRNA Details
Gene Symbol: Stxbp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053637.2 | NP_446089.1 | 3, 4 | 297 | ||
GenBank mRNA | AB046544.1 | - | 3, 4 | 298 | ||
BC098660.1 | - | 3, 4 | 225 | 7386801 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Stxbp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053637.2 | NP_446089.1 | 3, 4 | 297 | ||
GenBank mRNA | AB046544.1 | - | 3, 4 | 298 | ||
BC098660.1 | - | 3, 4 | 225 | 7386801 |