siRNA Details
| Gene Symbol: Asah2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053646.1 | NP_446098.1 | 4, 5 | 523 | ||
| XM_006231261.2 | XP_006231323.1 | 7, 8 | 881 | |||
| XM_006231263.1 | XP_006231325.1 | 6, 7 | 1459 | |||
| XM_006231267.2 | XP_006231329.1 | 6, 7 | 1465 | |||
| XM_006231268.2 | XP_006231330.1 | 6, 7 | 1462 | |||
| XM_006231270.1 | XP_006231332.1 | 6, 7 | 1462 | |||
| XM_008760319.1 | XP_008758541.1 | 9, 10 | 3301 | |||
| XM_008760320.1 | XP_008758542.1 | 6, 7 | 742 | |||
| GenBank mRNA | AB057433.1 | - | 4, 5 | 523 | ||

Asah2
Genomic Map