siRNA Details
| Gene Symbol: Vamp7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053531.1 | NP_445983.1 | 5 | 466 | ||
| XM_006249016.2 | XP_006249078.1 | 6 | 619 | |||
| GenBank mRNA | AF281632.1 | - | 5 | 466 | ||
| FQ228923.1 | - | 6 | 579 | |||
| FQ230649.1 | - | 5 | 390 | |||

Vamp7
Genomic Map