siRNA Details
| Gene Symbol: Tpra1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053534.2 | NP_445986.2 | 10 | 885 | ||
| XM_006236866.2 | XP_006236928.1 | 9 | 2124 | |||
| XM_006236868.2 | XP_006236930.1 | 9 | 925 | |||
| GenBank mRNA | AF292116.1 | - | 8 | 691 | ||
| FQ228747.1 | - | 10 | 960 | |||

Tpra1
Genomic Map