siRNA Details
| Gene Symbol: BMI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005180.8 | NP_005171.4 | 4, 5 | 767 | ||
| GenBank mRNA | BC011652.2 | - | 4, 5 | 766 | 4138748 | |
| BT019450.1 | - | 3, 4 | 261 | |||
| CR456911.1 | - | 3, 4 | 261 | |||
| L13689.1 | - | 4, 5 | 740 | |||
| Gene Symbol: COMMD3-BMI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001204062.1 | NP_001190991.1 | 8, 9 | 725 | ||
| GenBank mRNA | AK313235.1 | - | 8, 9 | 700 | ||

BMI1
Genomic Map