siRNA Details
| Gene Symbol: Gna15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053542.1 | NP_445994.1 | 6 | 919 | ||
| XM_006241021.2 | XP_006241083.1 | 6 | 1291 | |||
| GenBank mRNA | AB015308.1 | - | 6 | 994 | ||
| FQ232740.1 | - | 1 | 27 | |||
| FQ234321.1 | - | 1 | 27 | |||

Gna15
Genomic Map