siRNA Details
| Gene Symbol: Opn1mw | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053548.1 | NP_446000.1 | 2 | 381 | ||
| GenBank mRNA | AF031528.1 | - | 1 | 255 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Opn1mw| Gene Symbol: Opn1mw | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053548.1 | NP_446000.1 | 2 | 381 | ||
| GenBank mRNA | AF031528.1 | - | 1 | 255 | ||
Set Membership: |
|