siRNA Details
| Gene Symbol: Trpc6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053559.1 | NP_446011.1 | 2 | 695 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_001282087, NM_013838, NM_001282086, BC068310, AK137447, and others.

Trpc6
Genomic Map