siRNA Details
| Gene Symbol: Tinagl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053582.2 | NP_446034.1 | 8 | 1065 | ||
| XM_006238977.2 | XP_006239039.1 | 9 | 1157 | |||
| XM_006238978.2 | XP_006239040.1 | 9 | 1154 | |||
| GenBank mRNA | AB050717.1 | - | 8 | 1066 | ||
| BC097299.1 | - | 9 | 1124 | 7445067 | ||

Tinagl1
Genomic Map