siRNA Details
| Gene Symbol: Madd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053585.1 | NP_446037.1 | 8 | 1575 | ||
| GenBank mRNA | U72995.1 | - | 8 | 1575 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Madd| Gene Symbol: Madd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053585.1 | NP_446037.1 | 8 | 1575 | ||
| GenBank mRNA | U72995.1 | - | 8 | 1575 | ||