siRNA Details
| Gene Symbol: Bbs2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053618.1 | NP_446070.1 | 8 | 940 | ||
| XM_006255131.2 | XP_006255193.1 | 8 | 1099 | |||
| XM_008772263.1 | XP_008770485.1 | 8 | 867 | |||
| GenBank mRNA | AF342738.1 | - | 8 | 940 | ||
| FQ222286.1 | - | 3 | 157 | |||

Bbs2
Genomic Map