siRNA Details
| Gene Symbol: Cdc42bpb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053620.3 | NP_446072.2 | 8 | 1098 | ||
| XM_006240562.2 | XP_006240624.1 | 8 | 1247 | |||
| GenBank mRNA | AF021936.1 | - | 8 | 1058 | ||
| AY277590.1 | - | 8 | 1098 | |||

Cdc42bpb
Genomic Map