siRNA Details
| Gene Symbol: Sc5d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053642.2 | NP_446094.1 | 6 | 886 | ||
| GenBank mRNA | AB052846.1 | - | 5 | 864 | ||
| BC081704.1 | - | 6 | 886 | 7131154 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sc5d| Gene Symbol: Sc5d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053642.2 | NP_446094.1 | 6 | 886 | ||
| GenBank mRNA | AB052846.1 | - | 5 | 864 | ||
| BC081704.1 | - | 6 | 886 | 7131154 | ||