siRNA Details
| Gene Symbol: Tmf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053671.2 | NP_446123.2 | 1, 2 | 227 | ||
| XM_008763143.1 | XP_008761365.1 | 1, 2 | 1749 | |||
| GenBank mRNA | AF107843.1 | - | 1, 2 | 150 | ||
| BF415257.1 | - | 1, 2 | 227 | |||

Tmf1
Genomic Map