siRNA Details
| Gene Symbol: Dffa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053679.2 | NP_446131.2 | 1, 2 | 204 | ||
| XM_006239362.2 | XP_006239424.1 | 1, 2 | 225 | |||
| GenBank mRNA | AF136601.1 | - | 1, 2 | 189 | ||
| BC100067.1 | - | 1, 2 | 204 | 7389030 | ||

Dffa
Genomic Map